Learn More
Haldenwanger™ Porcelain Hirsch Funnel
Product Code. 10485821
Change view
Click to view available options
Description:
Size: 00
Size: 000
Diameter (Metric) Plate:
15mm
27mm
Diameter (Metric)Top:
52mm
72mm
Height (Metric):
74 mm
102 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Diameter (Metric) Plate Diameter (Metric)Top Height (Metric) Unit Size
10485821 Size: 00 27mm 72mm 102 mm Each
10643371 Size: 000 15mm 52mm 74 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10485821 Supplier Haldenwanger™ Supplier No. L 126/00
  • ONLINE EXCLUSIVE PRICE
    54.10€
    51.89€ / pezzi 1
    Risparmia 2.21€
    4% Sconto
Spedizione stimata: 23-11-2026
Designed with glazed porcelain for excellent thermal and chemical resistance. Haldenwanger™ Porcelain Hirsch Funnels are an excellent choice for quick filtration.
  • Sloped sides
  • Use for filtering small quantities
  • Perforated fixed filter plate
Specifications
Diameter (Metric)Top 72mm
Material Porcelain
Height (Metric) 102 mm
Description Size: 00
Diameter (Metric) Plate 27mm
Quantity 1Pack
Product Type Hirsch Funnel