Learn More
Thermo Scientific™ POROS™ 50 D RoboColumn™ Pre-packed columns
Codice prodotto. 18446698 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Capacità della colonna:
200 μL
600 μL
Dimensione della confezione:
200µL
600µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Capacità della colonna unitSize
18446698 200 μL 200µL
18406488 600 μL 600µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 18446698 Fornitore Thermo Scientific™ N. del fornitore A30723
Please to purchase this item. Need a web account? Register with us today!
Thermo Scientific™ RoboColumn™ chromatography products are small columns provided in 8-column strips.
RoboColumn™ chromatography products are small columns provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms. These particular RoboColumns contain POROS D50 resin, a 50-micron weak anion-exchange resin suitable for low-pressure applications, including large-scale downstream bioprocessing.
Features of the resin:
  • Higher productivity—through high throughput and high dynamic capacity associated with high performance chromatography
  • Chemical stability—allows aggressive cleaning and sanitization
  • Enhanced biomolecule access—provided via large pores (500-10,000 Å)
  • Polystyrenedivinylbenzene particles—yield a robust, easily packable matrix
  • Better separation methods in less time—the speed of high performance chromatography reduces weeks of experimentation to only a few hours, leaving more time to explore all the variables of your separation
RoboColumn™ is a registered trademark of ATOLL GmbH.
Specifica
Content And Storage Store at 2°C to 8°C or at room temperature.
Capacità della colonna 200 μL
Portata 16 to 1000 cm/hr.
Quantità 200 μL