missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ GoPure™ AAVX Column

Codice prodotto. 32044809 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Diametro (sistema metrico):
2.5 cm
5 cm
8 cm
10 cm
Lunghezza (sistema metrico):
5 cm
10 cm
Volume letto:
25 mL
50 mL
100 mL
200 mL
250 mL
390 mL
500 mL
785 mL
Dimensione della confezione:
Pezzo
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Diametro (sistema metrico) Lunghezza (sistema metrico) Volume letto unitSize
32044809 2.5 cm 5 cm 25 mL Pezzo
32044714 2.5 cm 10 cm 50 mL Pezzo
32044605 5 cm 5 cm 100 mL Pezzo
32044584 5 cm 10 cm 200 mL Pezzo
32045166 8 cm 5 cm 250 mL Pezzo
32045206 8 cm 10 cm 500 mL Pezzo
32045363 10 cm 5 cm 390 mL Pezzo
32045529 10 cm 10 cm 785 mL Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 32044809 Fornitore Thermo Scientific™ N. del fornitore A65878
 Articolo soggetto a restrizioni

Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.

Richiedi qui il prezzo per questo articolo.

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

POROS GoPure AAVX chromatography columns are ready-to-use and designed specifically for use in clinical and commercial manufacturing. POROS GoPure AAVX columns are prepared and packed in an ISO Class 7 environment where all product contact materials meet USP <88> Class VI standards.

POROS GoPure AAVX chromatography columns are ready-to-use and designed specifically for use in clinical and commercial manufacturing. POROS GoPure AAVX columns are prepared and packed in an ISO Class 7 environment where all product contact materials meet USP <88> Class VI standards. Each column is accompanied by column qualification documentation including bioburden and endotoxin. Packed with POROS™ CaptureSelect™ AAVX affinity resin, POROS GoPure AAVX columns offer high specificity and capacity for a broad range of AAV viral vector serotypes over a wide range of flow rates.

  • Pressure rating (bar/psi): 4/58
  • Operating temperature: 2°C to 30°C
  • Bed support size: 12 μm
  • Inlet/outlet internal diameter: 2.00 mm (2.5 cm), 3.45 mm (5 cm), 4.57 mm (8 cm), 6.35 mm (10 cm)
  • Inlet/outlet port connections: 3/4 inch mini tri-clamps
  • Includes a 30 mL resin side sample for incoming QC

Features of POROS CaptureSelect AAVX resin include one-step AAV purification from crude material with high purity, with high specificity and capacity, improving yield by reducing process steps. Free of animal-origin components, the CaptureSelect AAVX ligand is a 12 to 15 kDa single-domain antibody fragment produced in the yeast Saccharomyces cerevisiae.

Specifica

Volume letto 25 mL
Diametro (sistema metrico) 2.5 cm
Lunghezza (sistema metrico) 5 cm
Quantità 1 Ea.
Lunghezza (sistema imperiale) 2 in.

For research and development use and further manufacturing in support of FDA-regulated end uses. Not for diagnostic use or direct administration to human or animals.

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.