Learn More
Esculape Laboratoire™ Eyewash Bottles for First Aid
Product Code. 11769102
Change view
Click to view available options
Volume (Metric):
200 mL
500 mL
Composition:
Sterile Isotonic Solution
Sterile Ophthalmic Solution
Includes:
200 mL Blue Neutral pH Solution, 50 0mL Bottle Green Sterile Ophthalmic Solution
Cap
Cap, Storage Case, Belt Loop w/ Carrying Case
Cap, Storage, Carrying Case
Quantity:
10 Pk.
12 Pk.
Ea.
Unit Size:
Each
Pack of 10
Pack of 12
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Composition Includes Quantity Unit Size
11769102 200 mL Sterile Isotonic Solution Cap, Storage, Carrying Case Ea. Each
13588320 500 mL Sterile Ophthalmic Solution Cap 12 Pk. Each
11749522 500 mL Sterile Ophthalmic Solution Cap 12 Pk. Pack of 12
11962875 200 mL Sterile Isotonic Solution Cap, Storage Case, Belt Loop w/ Carrying Case Ea. Each
11759522 200 mL Sterile Ophthalmic Solution 200 mL Blue Neutral pH Solution, 50 0mL Bottle Green Sterile Ophthalmic Solution Ea. Each
11997625 200 mL Sterile Ophthalmic Solution Cap Ea. Each
11891553 200 mL Sterile Ophthalmic Solution Cap 10 Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11769102 Supplier Esculape Laboratoire™ Supplier No. 8205250
  • ONLINE EXCLUSIVE PRICE
    57.02€
    27.11€ / pezzi 1
    Risparmia 29.91€
    52% Sconto
 Restricted Item
Spedizione stimata: 03-12-2026
Use Esculape™ Eyewash Bottles for First Aid has a carrying case with a belt loop for portabilitiy.
  • Storage case
  • Contain sterile isotonic solution
  • Ergonomic
Specifications
Product Type Eyewash Bottle
Composition Sterile Isotonic Solution
For Use With (Application) Portable Eyewash
Includes Cap, Storage, Carrying Case
Quantity Ea.
Sterility Sterile
Volume (Metric) 200 mL