Learn More
Hanna Instruments™ Portable Lux Meter
Product Code. 10542284
Change view
Click to view available options
Range (Lux):
0.001 to 1.999
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range (Lux) Unit Size
10542284 0.001 to 1.999 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10542284 Supplier Hanna Instruments™ Supplier No. HI97500
  • ONLINE EXCLUSIVE PRICE
    424.94€
    294.81€ / pezzi 1
    Risparmia 130.13€
    30% Sconto
View all promotions
Spedizione stimata: 18-02-2027
Specifications
Type Portable Lux Meter
Accuracy ±6% of reading ±2 digits
Height (Metric) 45 mm
Length (Metric) 164 mm
Range (Lux) 0.001 to 1.999
Weight (Metric) 180 g
Width (Metric) 76 mm