Learn More
Brady™ Post and Cement Bases
Product Code. 11791306 Shop All Brady Products
Change view
Click to view available options
Color:
Black and Yellow Striped
Red
Red on White
White
Unit Size:
1 pack
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11791306 Red 1 pack
11721316 Red on White 1 pack
11711316 Black and Yellow Striped 1 pack
11741316 White 1 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11791306 Supplier Brady™ Supplier No. 292234
  • ONLINE EXCLUSIVE PRICE
    160.00€
    143.29€ / 1 confezioni
    Risparmia 16.71€
    10% Sconto
Spedizione stimata: 04-12-2026
These Post and Cement Bases are ideal for controlling and directing foot traffic. Suitable for warehouses, factories, offices etc. Chains and ribbons can be attached to the sturdy 900 mm posts.
  • Ideal for controlling and directing foot traffic.
  • Suitable for warehouses, factories, offices etc.
  • Chains and ribbons can be attached to the sturdy 900 mm posts.
  • Includes Post and Cement Base.
Specifications
Alternate Part Number BRADYPOST & CEMENT BASE RED PK4
Diameter (Metric) Outer 40 mm
Height (English) 35.43 in.
Height (Metric) 900 mm
Packaging Quantity 4 Pk.
Color Red
Type Post with Cement Base