Learn More
Potassium Trimethylacetate, TRC
Specifica
Specifica
| Materiale o nome chimico | Potassium Trimethylacetate |
| CAS | 19455-23-3 |
| Formula molecolare | C5 H9 O2 . K |
| Quantità | 5 g |
| Sinonimo | Propanoic acid','2,2-dimethyl-','potassium salt (1:1) (ACI), Pivalic acid','potassium salt (8CI), Propanoic acid','2,2-dimethyl-','potassium salt (9CI), 2,2-Dimethylpropanoic acid potassium salt, Potassium 2,2-dimethylpropanoate, Potassium pivalate, Potassium tert-butylcarboxylate, Potassium trimethylacetate, 2,2-Dimethylpropionic acid potassium salt |
| InChI Formula | InChI=1S/C5H10O2.K/c1-5(2,3)4(6)7;/h1-3H3,(H,6,7);/q;+1/p-1 |
| SMILES | [K+].CC(C)(C)C(=O)[O-] |
| IUPAC Name | potassium;2,2-dimethylpropanoate |
| Molecular Weight (g/mol) | 140.222 |
| Peso formulazione | 140.024 |