Learn More
UVP Power supply Pen-Ray PS9 RMS 2400 Vac lamp current 48mA 220-240V 50/60Hz for use with Pen-Ray lamp UVP
Product Code. 11828681
Change view
Click to view available options
For Use With (Equipment):
Pen-Ray Lamp UVP
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11828681 Pen-Ray Lamp UVP Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11828681 Supplier UVP Supplier No. 99003302
  • ONLINE EXCLUSIVE PRICE
    856.26€
    597.49€ / pezzi 1
    Risparmia 258.77€
    30% Sconto
Spedizione stimata: 11-01-2027