missing translation for 'onlineSavingsMsg'
Learn More

PPAP2A Recombinant Protein, Abnova

Marca:  Abnova H00008611-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16162662

  • 302.00€ / 10µg


Spedizione stimata
27-11-2020
Aggiungi al carrello

Descrizione

Descrizione

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifica

Specifica

AAH39847
Solution
8611
PPAP2A (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PPAP2A
Human
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
56.98
8
Glutathione Sepharose 4 Fast Flow
10 ug
MFDKTRLPYVALDVLCVLLAGLPFAILTSRHTPFQRGVFCNDESIKYPYKEDTIPYALLGGIIIPFSIIVIILGETLSVYCNLLHSNSFIRNNYIATIYKAIGTFLFGAAASQSLTDIAKYSIGRLRPHFLDVCDPDWSKINCSDGYIEYYICRGNAERVKEGRLSFYSGHSSFSMYCMLFVALYLQARMKGDWARLLRPTLQFGLVAVSIYVGLSRVSDYKHHWSDVLTGLIQGALVAILVAVYVSDFFKERTSFKERKEEDSHTTLHETPTTGNHYPSNHQP
LLP1a|LPP1|PAP-2a|PAP2|PAP2a2|PAP2alpha2|PAPalpha1
PPAP2A
Wheat Germ (in vitro)
GST
Videos
SDS
Documenti