missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPARA Mouse anti-Human, Clone: 1G1E10, Proteintech
Mouse Monoclonal Antibody
140.00€ - 399.00€
Specifica
Antigene | PPARA |
---|---|
Clone | 1G1E10 |
Concentrazione | 1 mg/mL |
Applicazioni | Western Blot |
Classificazione | Monoclonal |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
16809065
|
Proteintech
66826-1-IG-20UL |
20 μL |
140.00€
20µL |
Spedizione stimata: 13-05-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
16899055
|
Proteintech
66826-1-IG-150UL |
150 μL |
399.00€
150µL |
Spedizione stimata: 13-05-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Descrizione
Peroxisome proliferators are non-genotoxic carcinogens which are purported to exert their effect on cells through their interaction with members of the nuclear hormone receptor family termed peroxisome proliferator activated receptors (PPAR′s). Nuclear hormone receptors are ligand-dependent intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences following activation by the appropriate ligand. Studies indicate that PPAR′s are activated by peroxisome proliferators such as clofibric acid, nafenopin, and WY-14,643, as well as by some fatty acids. It has also been shown that PPAR′s can induce transcription of acyl coenzyme A oxidase & cytochrome P450 (CYP450) A6 through interaction with specific response elements. PPAR, like several other nuclear hormone receptors, heterodimerizes with retinoid X receptor (RXR) alpha.Specifica
PPARA | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
hPPAR, NR1C1, PPAR, PPAR alpha, PPARA | |
PPARA | |
IgG1 | |
Protein G | |
Antibody |
1G1E10 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q07869 | |
5465 | |
PPARA Fusion Protein Ag7896 | |
Primary | |
-20°C | |
PPARA |