Learn More
Periplakin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA563784
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: AKEERINKLH SEGDQLLAAE HPGRNSIEAH MEAVHADWKE YLNLLICEES HLKYMEDYHQ FHEDVKDAQE LLRKVDSDLN QKYGPDFKDR YQIELLL Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
Periplakin Polyclonal antibody specifically detects Periplakin in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifica
Periplakin | |
Polyclonal | |
Unconjugated | |
Ppl | |
190 kDa paraneoplastic pemphigus antigen; 195 kDa cornified envelope precursor protein; 195kDa protein; AW553870; KIAA0568; MGC134872; Periplakin; Ppl | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5493 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60437 | |
Ppl | |
Recombinant protein corresponding to Human PPL | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only