missing translation for 'onlineSavingsMsg'
Learn More

PR48 Antibody [Janelia Fluor« 549], Novus Biologicals Biologicals™

Codice prodotto. 30516296 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.1 mL
Dimensione della confezione:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30516296 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30516296 Fornitore Novus Biologicals N. del fornitore NBP338554JF549

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

PR48 Polyclonal antibody specifically detects PR48 in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Especificaciones

Antigene PR48
Applicazioni ELISA, Western Blot
Classificazione Polyclonal
Coniugato Janelia Fluor 549
Formulazione 50mM Sodium Borate
Alias gene FLJ60425, NYREN8, NY-REN-8 antigen, PP2A subunit B isoform PR48, PP2A, subunit B, PR48 isoform, PPP2R3L, PPP2R3LY, PR48, protein phosphatase 2 (formerly 2A), regulatory subunit B'', beta, protein phosphatase 2, regulatory subunit B'', beta, Protein phosphatase 2A 48 kDa regulatory subunit, serine/threonine protein phosphatase 2A, 48kDa regulatory subunit B, serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit beta
Specie ospite Rabbit
Immunogeno Recombinant fusion protein containing a sequence corresponding to amino acids 356-575 of human PR48 (NP_037371.2).,, Sequence:, FSGAVTRGRKVQKEGKISYADFVWFLISEEDKKTPTSIEYWFRCMDLDGDGALSMFELEYFYEEQCRRLDSMAIEALPFQDCLCQMLDLVKPRTEGKITLQDLKRCKLANVFFDTFFNIEKYLDHEQKEQISLLRDGDSGGPELSDWEKYAAEEYDILVAEETAGEPWEDGFEAELSPVEQKLSALRSPLAQRPFFEAPSPLGAVDLYEYACGDEDLEPL
Metodo di purificazione Affinity purified
Quantità 0.1 mL
Status giuridico RUO
Disciplina di ricerca Cell Cycle and Replication
Primario o secondario Primary
ID gene (immissione) 28227
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Forma Purified
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.