Learn More
Pradigastat, MedChemExpress
Codice prodotto. 16493067
Encompass_Preferred
Cambia vista
Click to view available options
Quantità:
5 mg
10 mg
50 mg
Dimensione della confezione:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
16493067 50 mg 50mg
16487018 10 mg 10mg
16429567 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Codice prodotto. 16493067 Fornitore Medchem Express N. del fornitore HY16278
MedChemExpress Pradigastat (LCQ-908) is a potent, selective and orally active diacylglycerol acyltransferase 1 (DGAT1) inhibitor. Pradigastat has anti-obesity and anti-diabetic effects.
Pradigastat (LCQ-908) is a potent, selective and orally active diacylglycerol acyltransferase 1 (DGAT1) inhibitor. Pradigastat has anti-obesity and anti-diabetic effects.
Specifica
Materiale o nome chimico Pradigastat
CAS 956136-95-1
Colore Light Yellow
Note sul grado di purezza Research
Formula molecolare C25H24F3N3O2
Quantità 50 mg
Sinonimo LCQ-908
Informazioni di solubilità DMSO : 16.67 mg/mL (36.60 mM; ultrasonic and warming and heat to 60°C) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(O)C[C@H]1CC[C@H](C2=CC=C(C3=NC=C(NC4=CC=C(C(F)(F)F)N=C4)C=C3)C=C2)CC1
Molecular Weight (g/mol) 455.47
Peso formulazione 455.47
Percent Purity 98.0%
Grado Research
Forma fisica Powder
Da utilizzare con (applicazione) Metabolism-sugar/lipid metabolism
Research purposes only