Learn More
Tocris Bioscience™ Prasugrel
Irreversible P2Y12 antagonist; antiplatelet and orally active
Marca: Tocris Bioscience™ 6317/10
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Prasugrel is a irreversible P2Y12 antagonist; metabolised to active metabolite R-99224 (IC50 = 45 μM) irreversibly binds platelet P2Y12 receptors, inhibiting platelet activation. Antiplatelet and antithrombotic. Orally active and active in vivo. Note - inactive in vitro.Specifica
| 2-[2-(Acetyloxy)-6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl]-1-cyclopropyl-2-(2-fluorophenyl)ethanone | |
| 150322-43-3 | |
| >98% | |
| 10 mg |