missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Precise™ 8%, Tris-Glycine, 7.0 mm, 12-well

Codice prodotto. 11834594 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Quantità:
10/Pk.
Dimensione della confezione:
pezzi 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
11834594 10/Pk. pezzi 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Normativo: Questo prodotto contiene una sostanza elencata nell'allegato XVII del regolamento REACH (CE) n. 1907/2006. Tutti i clienti devono compilare una dichiarazione prima della consegna.
Codice prodotto. 11834594 Fornitore Thermo Scientific™ N. del fornitore 25250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Normativo: Questo prodotto contiene una sostanza elencata nell'allegato XVII del regolamento REACH (CE) n. 1907/2006. Tutti i clienti devono compilare una dichiarazione prima della consegna.

Separation Range: 45 to 205 kd; Separation Type: Denaturing, Native; For Use With: Novex XCell I and II; Novex XCell II Surelock; IBI Scientific Universal Protein Systems

Thermo Scientific™ Precise Tris-Glycine Gels are long shelf-life, fast run-time polyacrylamide gels for SDS-PAGE that use the popular Laemmli buffer system and commercial gel tanks to provide outstanding protein resolution and separation.

Precise Tris-Glycine Gels are cast in durable 10 x 10cm plastic cassettes to fit in Novex™ XCell gel tanks. The gels are individually packaged in easy-to-open plastic pouches and are ready for electrophoresis with no comb or tape to remove. Following electrophoresis, the easy-open cassette design allows gels to be removed damage-free without special tools. Precise Gels in the 10 x 10cm size are available in gradient (4 to 20% and 8 to 16%) and fixed (8%, 10% and 12%) concentrations and in 10-well and 12-well formats.

Highlights:

  • High-performance – Tris-glycine running buffer produces excellent separation and high-resolution protein bands

  • Long shelf-life – up to 12-month stability ensures consistent performance for the duration of each package of gels

  • Fast – 60-minute run-time provides results quickly; transfers efficiently to nitrocellulose and PVDF membranes; stains quickly with coomassie and silver stains

  • Compatible – these 10 x 10cm cassettes fit the Novex™ XCell and a other similar tanks

  • Easy to load – sample wells reinforced with plastic to eliminate damage when loading and well-to-well contamination

Specifications:

  • Cassette Dimensions: 10cm × 10cm × 7mm

  • Gel Dimensions: 8cm × 8.8cm × 1mm

  • Storage Conditions: 4°C

  • Shelf Life: 12 months from date of purchase

  • Stacking Gel: 4%

  • Buffer System in Gel: Tris-HCl, pH 7

  • SDS: None

  • Required Running Buffer: Tris-glycine-SDS

  • Recommended Sample Buffer: Tris-HCl-SDS or LDS-SDS

  • Gel Tank CompatibilityCompatibility (10cm x 10cm x 7mm): Novex XCell I and II; Novex XCell II™ Surelock™; IBI Scientific Universal Protein Systems; For gels that are compatible with other tanks, see our 10 x 8cm Gels

TRUSTED_SUSTAINABILITY

Specifica

Descrizione 8% Tris-Glycine Gels, 10 x 10cm, 12-well
Gel Percentage 8%
Gel Thickness 0.7 cm
Gel Type Tris-Glycine
Separation Range 45 to 205 kDa
Separation Type Denaturing, Native
Pozzetti 12-well
Formulazione Precast 8% polyacrylamide gels for SDS-PAGE
Quantità 10/Pk.
Sufficiente per 12 samples per gel, 30uL capacity per well
Design pozzetto Standard
Lunghezza (sistema metrico) 10 cm
Larghezza (sistema metrico) 10 cm
Product Type Homogeneous Page
Da utilizzare con (apparecchiatura) Novex XCell I and II, Novex XCell II Surelock, IBI Scientific Universal Protein Systems
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.