Learn More
MP BIOMEDICALS Precision Tissue Wipers
Codice prodotto. 30928576 Sfoglia Tutto MP Biomedicals Prodotti
Cambia vista
Click to view available options
Product Type:
Low-lint Wiper
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Product Type
30928576 Low-lint Wiper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30928576 Fornitore MP BIOMEDICALS N. del fornitore 07DP1030
  • ONLINE EXCLUSIVE PRICE
    18.75€
    17.85€ / pezzi 280
    Risparmia 0.90€
    4% Sconto
Visualizza tutte le promozioni
Spedizione stimata: 13-11-2026
Low-lint wipers for delicate applications.
  • Kim Wipes designed for delicate laboratory and manufacturing tasks requiring minimal lint and particle generation.
  • Ideal for cleaning sensitive surfaces, instruments, and optical equipment without causing damage.
  • Supplied for research or further manufacturing use only; not intended for direct human use.
  • Durable and absorbent, providing efficient wiping and contamination control in precise laboratory environments.
Specifica
Product Type Low-lint Wiper
Materiale Tissue