Learn More
Prednisone, 98%, Thermo Scientific Chemicals
Sfoglia Tutto Thermo Scientific Chemicals ProdottiDescrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Prednisone is a synthetic glucocorticoid with anti-inflammatory and immunosuppressive activities
• It shows antineoplastic and adrenergic activities against hormone-sensitive cancer cells
• This compound binds to its receptors and enters the nucleus activating specific nuclear receptors, resulting in an altered gene expression and inhibiting proinflammatory cytokine production
Applications
• Prednisone can reduce circulating lymphocytes, promote cell differentiation, and stimulate apoptosis in sensitive tumor cell lines
• It is considered a prodrug that is converted by the liver into prednisolone which is the active steroid compound
• It is also used in some studies to investigate the effects of daily administration of MIDD0301, on the animal and organs weights and hematological profile
Specifica
Specifica
| Materiale o nome chimico | Prednisone |
| Intervallo percentuale saggio | 98% |
| Imballaggio | Bottiglia in vetro |
| Quantità | 5 g |
| Peso formulazione | 358.43 |
RUO – Research Use Only
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.