Descrizione
- Include annual maintenance with:
- Check of installation environment and electrical/gas supply
- Inspection of equipment-specific sensors, doors/lids and key components
- Validation and adjustment of operating parameters
- Equipment-specific differences:
- Multipurpose Centrifuges:
- Housing and rotor cleaning
- Checks of condensation tray, lid latch, display, keypad, stability, rotor/imbalance detection, temperature and speed sensors
- New Brunswick S41i:
- CO2 and auto-zero filter replacement
- Checks of shaker platform, drive belt, humidity tray and support seals
- CellXpert CS220:
- CO2 filter replacement; checks of shaker platform, drive belt and rH sensor
- CellXpert C 170 (i):
- Gas and auto-zero filter replacement; checks of door, alarm, humidity tray, wiring, PCB, display and controller
- Biological Shakers:
- Door/lid hardware checks
- Lubrication of shaker mechanism
- Checks of drive motor and belt
- Condenser cleaning and inspection
- Multipurpose Centrifuges:
Specifica
Specifica
| Product Type | Service Plan |
| Service Offering | Multipurpose Centrifuges with Refrigeration |