Learn More
Asecos™ Presence Sensor ION-ULTRA-90
Product Code. 31789612 Shop All asecos Products
Change view
Click to view available options
For Use With (Application):
Delays auto-close
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
31789612 Delays auto-close Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31789612 Supplier Asecos™ Supplier No. 42169
 Restricted Item
Spedizione stimata: 22-12-2026
Optional sensor electronics accessory for the ION-ULTRA-90 model of the asecos ION-LINE safety storage cabinet
  • With the presence sensor, doors remain open automatically as long as it detects people within the swing range.
  • For maximum ease of use when loading and unloading the cabinet.
  • It works with the cabinets built-in door safety system - preserving the automatic door closing and acoustic warning once the user leaves the detection zone.
Specifications
For Use With (Application) Delays auto-close
Weight (English) 2.20 lb.
Weight (Metric) 1 kg
Product Type Presence Sensor