Learn More
Proteintech Prestained Protein Marker
The PL00001 Regular Range Protein Marker is a ready to use three-color protein standard with 10 prestained proteins covering a wide range of molecular weights from 10 to 180 kDa.
Marca: Proteintech PL00001-100UL
Descrizione
- The PL00001 Prestained Protein Marker is designed for monitoring protein separation during SDS-polyacrylamide gel electrophoresis, verification of Western transfer efficiency on membranes, (PVDF, nylon, or nitrocellulose) and for estimating the size of proteins.
- Contents: Approximately 0.1~0.4 mg/mL of each protein in the buffer (20 mM Tris-phosphate (pH 7.5 at 25°C), 2 % SDS, 0.2 mM Dithiothreitol, 3.6 M Urea, and 15 % (v/v) Glycerol).