Learn More
JULABO PRESTO Process Systems
Codice prodotto. 17744869
Cambia vista
Click to view available options
Modello:
A38
A41
A70
W41
Dimensione della confezione:
Pezzo
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Modello Dimensione della confezione
17744869 W41 Pezzo
17714869 A38 Pezzo
17724869 A41 Pezzo
17734869 A70 Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Codice prodotto. 17744869 Fornitore JULABO N. del fornitore 9421411.N1
 Articolo soggetto a restrizioni
Spedizione stimata: 03-02-2027
The PRESTO systems are designed for precise temperature control as well as rapid temperature changes, making them ideal for reactor vessels, material stress tests, or temperature simulations.
  • Wide working temperature ranges without changing fluids
  • Ethernet interface
  • SD-Card slot
  • pace optimized design creates more room directly next to the units
  • Connections for USB, Ethernet, RS232, and Alarm output
Specifica
Temperature Range (Metric) -45°C to +250°C
Dimensioni (L x P x A) 29.5 x 13 x 26.4 in.
Modello W41
Tipo Process System