Descrizione
Prints each cassette in 10 seconds or less
Small footprint allows the printer to fit easily next to the grossing station
Print only the cassettes you need, right at the point of use Print barcodes on cassettes to automate downstream processes Eliminate manual handwriting processes
Small footprint allows the printer to fit easily next to the grossing station
Print only the cassettes you need, right at the point of use Print barcodes on cassettes to automate downstream processes Eliminate manual handwriting processes
Specifica
Specifica
| Descrizione | PrintMate AS 450 Cassette Printer |
| Capacità | 450 Cassettes |
| Include | PrintMate AS USB cable Power cable 6 cassette hoppers One roll of tape (installed) Epredia™ LabWriter™ software CD Operator guide |
| Requisiti elettrici | 100/240 VAC, 50/60 Hz, 150 VA (512 BTU/h) |
| Product Type | Cassette Printer |