Learn More
- Home
- Products
- Histology and Cytology
- Tissue Embedding
- Embedding Cassettes and Storage
- Epredia™ PrintMate™ Biopsy Cassettes
Epredia™ PrintMate™ Biopsy Cassettes
Codice prodotto. 16364026 Sfoglia Tutto Epredia Prodotti
Cambia vista
Click to view available options
Colore:
Aqua
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Quantità:
10 x 75 cassettes (750)/cs
4 x 250 cassettes (1000)/cs
Dimensione della confezione:
pezzi 1000
pezzi 750
34 product options available for selection
| Codice prodotto. | Colore | Quantità | Dimensione della confezione |
|---|---|---|---|
| 16364026 | Yellow | 10 x 75 cassettes (750)/cs | pezzi 750 |
| 15487604 | Aqua | - | pezzi 1000 |
| 16313986 | Beige | 4 x 250 cassettes (1000)/cs | pezzi 1000 |
| 15497604 | Blue | - | pezzi 1000 |
| 16232321 | Blue | 4 x 250 cassettes (1000)/cs | pezzi 1000 |
| 15407614 | Green | - | pezzi 1000 |
| 16323986 | Green | 4 x 250 cassettes (1000)/cs | pezzi 1000 |
| 15417614 | Gray | - | pezzi 1000 |
| 16333986 | Gray | 4 x 250 cassettes (1000)/cs | pezzi 1000 |
| 15427614 | Lilac | - | pezzi 1000 |
| 16343986 | Lavender | 4 x 250 cassettes (1000)/cs | pezzi 1000 |
| 15437614 | Orange | - | pezzi 1000 |
| 16353986 | Light Green | 4 x 250 cassettes (1000)/cs | pezzi 1000 |
| 15447614 | Pink | - | pezzi 1000 |
| 16363986 | Lilac | 4 x 250 cassettes (1000)/cs | pezzi 1000 |
| 15457614 | Tan | - | pezzi 1000 |
| 16373986 | Orange | 4 x 250 cassettes (1000)/cs | pezzi 1000 |
| 15467614 | White | - | pezzi 1000 |
| 16383986 | Pink | 4 x 250 cassettes (1000)/cs | pezzi 1000 |
| 15477614 | Yellow | - | pezzi 1000 |
| 16393986 | Tan | 4 x 250 cassettes (1000)/cs | pezzi 1000 |
| 16303996 | White | 4 x 250 cassettes (1000)/cs | pezzi 1000 |
| 16313996 | Yellow | 4 x 250 cassettes (1000)/cs | pezzi 1000 |
| 16354016 | Beige | 10 x 75 cassettes (750)/cs | pezzi 750 |
| 16364016 | Blue | 10 x 75 cassettes (750)/cs | pezzi 750 |
| 16374016 | Green | 10 x 75 cassettes (750)/cs | pezzi 750 |
| 16384016 | Gray | 10 x 75 cassettes (750)/cs | pezzi 750 |
| 16394016 | Lavender | 10 x 75 cassettes (750)/cs | pezzi 750 |
| 16304026 | Light Green | 10 x 75 cassettes (750)/cs | pezzi 750 |
| 16314026 | Lilac | 10 x 75 cassettes (750)/cs | pezzi 750 |
| 16324026 | Orange | 10 x 75 cassettes (750)/cs | pezzi 750 |
| 16334026 | Pink | 10 x 75 cassettes (750)/cs | pezzi 750 |
| 16344026 | Tan | 10 x 75 cassettes (750)/cs | pezzi 750 |
| 16354026 | White | 10 x 75 cassettes (750)/cs | pezzi 750 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 34 options available.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Artikelnummer.
16364026
Lieferant
Epredia™
Lieferanten-Nr.
B851110YW
-
102.00€
78.93€ / pezzi 750
Risparmia 23.07€
22% Sconto
Deve Accedi o registrarsi per acquistare questo articolo.
Articolo soggetto a restrizioni
In stock
Eseguire il login per visualizzare lo stock disponibile
Eseguire il login per visualizzare lo stock disponibile
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Designed for performance and user convenience, the PrintMate cassette range has been optimized for superior workflow and printing with the PrintMate Classic and PrintMate AS printing systems. The even, textured printing face provides the optimal surface for high-quality printing every time. For additional information, the right and left sides are textured for clear handwriting with suitable pens. The lid catches have been adjusted for ease of use with low force opening and closing, while still giving the security needed while processing.Available in tubes for automatic loading or loose for manual use, and in twelve colors for enhanced workflow identification.
Spezifikation
Spezifikation
| Stile pori | Small Square Pores |
| Chiusura inclusa | Separate lids included |
| Quantità | 10 x 75 cassettes (750)/cs |
| Tipo di chiusura | Reversible |
| Compatibilità con PrintMate | Yes |
| Colore | Yellow |
| Product Type | PrintMate Biopsy Cassette for PrintMate Printers |
Name des Produkts
Wählen Sie ein Thema