Learn More
PRMT5 Antibody (RAB-C136) - F(ab) - Azide and BSA Free, Novus Biologicals™
Codice prodotto. 18698034 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.025 mg
0.1 mg
Dimensione della confezione:
0.02mg
0.10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18698034 0.025 mg 0.02mg
18618454 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 18698034 Fornitore Novus Biologicals N. del fornitore NBP3201300.025mg
Human Monoclonal Antibody
PRMT5 Monoclonal antibody specifically detects PRMT5 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP assay (ChIP)
TRUSTED_SUSTAINABILITY
Specifica
Antigene PRMT5
Applicazioni ELISA, Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP Assay
Classificazione Monoclonal
Clone RAB-C136
Concentrazione 1 mg/mL
Coniugato Unconjugated
Formulazione PBS
Alias gene EC 2.1.1.-, EC 2.1.1.125, Histone-arginine N-methyltransferase PRMT5, HMT1 hnRNP methyltransferase-like 5,72 kDa ICln-binding protein, HRMT1L5, IBP72, Jak-binding protein 1, JBP1, protein arginine methyltransferase 5, protein arginine N-methyltransferase 5, Shk1 kinase-binding protein 1 homolog, SKB1, skb1 (S. pombe) homolog, SKB1 homolog (S. pombe), SKB1HsSKB1 homolog
Specie ospite Human
Immunogeno This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRMT5 protein under non-denaturing conditions.
Metodo di purificazione Affinity purified
Quantità 0.025 mg
Status giuridico RUO
Disciplina di ricerca Cell Cycle and Replication, Chromatin Research, Circadian Rhythm
Primario o secondario Primary
ID gene (immissione) 10419
Target Species Human
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Forma Purified