Learn More
Memmert™ Process-Dependent Programmable Door Lock
Process-Dependent Programmable Door Lock
Marca: Memmert™ IPP2200ecoD43
Dettagli aggiuntivi : Peso : 0.01100kg
Descrizione
- Process-Dependent Programmable Door Lock
- Accessory
Specifica
IPP2200eco | |
Programmable Door Lock | |
Three interlocking systems | |
Only for Units with TwinDISPLAY |