Learn More
Promega Sequencing Grade Modified Trypsin, Lyophilized
Sequencing Grade Trypsin has been manufactured to provide maximum specificity. Lysine residues in porcine trypsin have been modified by reductive methylation, yielding a highly active and stable molecule that is extremely resistant to autolytic digestion.
Marca: Promega V5111
Includes: 1 × 1mL Trypsin Resuspension Dilution Buffer, 5 × 20μg Sequencing Grade Modified Trypsin, Porcine
Descrizione
- Manufactured for Maximum Specificity and Stability
- Lysine residues modified by reductive methylation, yielding a highly active and stable trypsin
- Available lyophilized or frozen
- Referenced in thousands of papers
Specifica
| 0.1 mg/mL | |
| 7.7 | |
| 50mM Acetic Acid Resuspension Buffer | |
| 5 x 20 μg |
| White | |
| 'Hydrolyzing the Peptide at the Carboxyl side of Lysine and Arginine residues | |
| -30°C to -10°C |