Learn More
Promega Sequencing Grade Modified Trypsin, Lyophilized
Sequencing Grade Trypsin has been manufactured to provide maximum specificity. Lysine residues in porcine trypsin have been modified by reductive methylation, yielding a highly active and stable molecule that is extremely resistant to autolytic digestion.
Marca:  Promega V5111
Encompass_Preferred
Codice prodotto. 16563621
  • 172.80€ / 100µg
Includes: 1 × 1mL Trypsin Resuspension Dilution Buffer, 5 × 20μg Sequencing Grade Modified Trypsin, Porcine
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
  • Manufactured for Maximum Specificity and Stability
  • Lysine residues modified by reductive methylation, yielding a highly active and stable trypsin
  • Available lyophilized or frozen
  • Referenced in thousands of papers
Specifica
Specifica
0.1 mg/mL
7.7
50mM Acetic Acid Resuspension Buffer
5 x 20 μg
White
'Hydrolyzing the Peptide at the Carboxyl side of Lysine and Arginine residues
-30°C to -10°C