Learn More
Propargylamine, Hydrochloride, TRC
Product Code. 30421478
Change view
Click to view available options
Quantity:
2.5 g
5 g
10 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30421478 5 g -
30423258 10 g -
30421332 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30421478 Supplier Toronto Research Chemicals Supplier No. TRCP7625255G
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 15430-52-1
Molecular Formula C3 H5 N . Cl H
Molecular Weight (g/mol) 91.54
IUPAC Name prop-2-yn-1-amine;hydrochloride
SMILES Cl.NCC#C
Specifications
Quantity 5 g
InChI Formula InChI=1S/C3H5N.ClH/c1-2-3-4;/h1H,3-4H2;1H
Formula Weight 91.0189
Chemical Name or Material Propargylamine, Hydrochloride