Learn More
Propiolic Acid, TRC
Specifica
Specifica
| Materiale o nome chimico | Propiolic Acid |
| CAS | 471-25-0 |
| Formula molecolare | C3H2O2 |
| Quantità | 25 g |
| Sinonimo | 2-Propyne-1-carboxylic Acid, Acetylenecarboxylic Acid, Acetylenemonocarboxylic Acid, Carboxyacetylene, NSC 16152, Propargylic Acid, Propynoic Acid |
| InChI Formula | InChI=1S/C3H2O2/c1-2-3(4)5/h1H,(H,4,5) |
| SMILES | OC(=O)C#C |
| IUPAC Name | prop-2-ynoic acid |
| Molecular Weight (g/mol) | 70.05 |
| Peso formulazione | 70.0055 |