Learn More
Invitrogen™ ProQuantum™ Software v1.1
Codice prodotto. 16595651
Cambia vista
Click to view available options
Quantità:
1 License Key
5 License Keys
Dimensione della confezione:
Pezzo
pezzi 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
16595651 1 License Key Pezzo
16505661 5 License Keys pezzi 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 16595651 Fornitore Invitrogen™ N. del fornitore A43594
  • ONLINE EXCLUSIVE PRICE
    1150.00€
    908.50€ / pezzi 1
    Risparmia 241.50€
    21% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 22-01-2027
Enables you to analyze ProQuantum high-sensitivity immunoassay results acquired from any qPCR instrument.
ProQuantum Software enables you to analyze ProQuantum high-sensitivity immunoassay results acquired from any qPCR instrument. The analysis features include standard curve qualification with built-in outlier detection, interpolation of unknown samples for quantitation, and group-wise statistical analysis. Step-by-step guidance is provided for preparation of an assay run, including setting up the standard curve and plate layout for 96- or 384-well plates.
• Convenient setup wizard to minimize errors―automatic calculations for standard curve serial dilution setup
• Built-in statistical comparison―easily compare controls vs treatment groups
• Export capabilities―view results in Excel and PDF, or conveniently formatted for GraphPad Prism
ProQuantum Software consists of two modules: Create Experiment and Import and View Results.
• Create Experiment allows you to set up a standard curve, create a plate layout, and assign descriptions and groups to individual samples wells
• Import and View Results allows you to import experimental data adjusted to fit the standard curve on a 4PL or 5PL weighted regression algorithm, interpolate unknown protein concentrations, view heat maps, perform group-wise comparisons, and export the data
ProQuantum Software is compatible with all ProQuantum immunoassay kits and with all Applied Biosystems qPCR instruments, including QuantStudio 3/5/6 Flex/7 Flex/12K Flex, StepOne, StepOnePlus, 7500, 7500 Fast, ViiA 7, or other compatible qPCR instruments.
The preferred operating system is Windows 7 or 10.
For more information, visit thermofisher.com/proquantum.
TRUSTED_SUSTAINABILITY
Specifica
Tipo ProQuantum Software v1.0
Da utilizzare con (apparecchiatura) 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, BioRad CFX96™, QuantStudio™, ViiA™ 7 System, StepOnePlus™ System, Roche LightCycler 480, Bio-Rad CFX384 Touch™, Bio-Rad CFX Connect™, StepOne™ System, Bio-Rad CFX96 Touch™
Linea di prodotti ProQuantum
Quantità 1 License Key
Content And Storage Includes 1 license key for 1 user
Software Data Analysis Software