Learn More
EKASTU Safety Protective Hood Universal
Codice prodotto. 10696275
Cambia vista
Click to view available options
Colore:
grigio
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Colore
10696275 grigio
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 10696275 Fornitore EKASTU Safety N. del fornitore 286714
 Articolo soggetto a restrizioni
Spedizione stimata: 27-01-2027
Protective hood offers neck and throat protection. Breathable and comfortable.
  • Constructed of very tough tissue
  • Versatile use
  • Durable and resistant
  • Washable
Specifica
Product Type circa. 70 grammi
Size universale
Colore grigio
Monouso no
Quantità un pezzo
Dimensioni (L x P x A) 22 x 11 x 2cm
Peso (sistema metrico) Approx. 70 g