Learn More
Tocris Bioscience™ Proteome Profiler Rat XL Cytokine Array
Descrizione
The Proteome Profiler Rat XL Cytokine Array contains 4 membranes-each spotted in duplicate with 79 different cytokine antibodies.
Specifica
Specifica
| Descrizione | Four membranes-each spotted in duplicate with 79 different cytokine antibodies |
| Product Type | Capture Antigen ELISA kit |
| Content And Storage | Array Buffer, Chemi Reagent, Detection Antibody Cocktail, Rat XL Cytokine Array, Rat XL Cytokine Array nitrocellulose membranes, Rectangular four-well multi-dish, Streptavidin-HRP, Transparency Overlay Template, Wash Buffer Concentrate (25X). Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
For Laboratory Research Use Only