missing translation for 'onlineSavingsMsg'
Learn More

PRPF39 Antibody - BSA Free, Novus Biologicals™

Codice prodotto. 30557057 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
100 μg
Dimensione della confezione:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30557057 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30557057 Fornitore Novus Biologicals N. del fornitore NBP346883

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Rabbit Polyclonal Antibody

PRPF39 Polyclonal antibody specifically detects PRPF39 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifica

Antigene PRPF39
Applicazioni ELISA, Immunohistochemistry, Western Blot
Classificazione Polyclonal
Coniugato Unconjugated
Diluizione Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:20-1:200
Formulazione PBS, 50% Glycerol, pH (7.3)
Alias gene FLJ11128, FLJ20666, FLJ45460, MGC149842, MGC149843, pre-mRNA-processing factor 39, PRP39 homolog, PRP39 pre-mRNA processing factor 39 homolog (S. cerevisiae), PRP39 pre-mRNA processing factor 39 homolog (yeast)
Specie ospite Rabbit
Immunogeno PRP39 pre-mRNA processing factor 39 homolog. (Uniprot# Q86UA1)
Metodo di purificazione Immunogen affinity purified
Quantità 100 μg
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 55015
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Forma Purified
Isotype IgG
Vedi altri risultati Mostra meno risultati
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.