missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PSMA4 (Human) Recombinant Protein
Human PSMA4 full-length ORF ( AAH05361, 1 a.a. - 261 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00005685-P01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
- Sequence: MSRRYDSRTTIFSPEGRLYQVEYAMEAIGHAGTCLGILANDGVLLAAERRNIHKLLDEVFFSEKIYKLNEDMACSVAGITSDANVLTNELRLIAQRYLLQYQEPIPCEQLVTALCDIKQAYTQFGGKRPFGVSLLYIGWDKHYGFQLYQSDPSGNYGGWKATCIGNNSAAAVSMLKQDYKEGEMTLKSALALAIKVLNKTMDVSKLSAEKVEIATLTRENGKTVIRVLKQKEVEQLIKKHEEEEAKAEREKKEKEQKEKDK
Specifica
AAH05361 | |
proteasome (prosome, macropain) subunit, alpha type, 4 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PSMA4 | |
GST |
5685 | |
Wheat germ expression system | |
25 μg | |
HC9, HsT17706, MGC111191, MGC12467, MGC24813, PSC9 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |