Learn More
PSMB1 Mouse, anti-Human, Unconjugated, Clone: 1C7C5, Proteintech
Mouse Monoclonal Antibody
Marca:  Proteintech 67847-1-IG-20UL
Dettagli aggiuntivi : Peso : 0.00020kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17231516
  • 129.00€ / 20µL
Spedizione stimata: 19-12-2022
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
PSMB1(Proteasome subunit beta type-1) is also named as PSC5 and belongs to the peptidase T1B family. The proteasome is a multicatalytic proteinase complex which is characterized by its ability to cleave peptides with Arg, Phe, Tyr, Leu, and Glu adjacent to the leaving group at neutral or slightly basic pH. The gene encodes a 241 amino acid protein with a 28 amino acid propeptide and two glycosylation sites.
Specifica
Specifica
PSMB1
Monoclonal
Unconjugated
20 μL
5689
Liquid
ELISA, Immunohistochemistry, Western Blot
1C7C5
Mouse