missing translation for 'onlineSavingsMsg'
Learn More

PSMB7 Mouse anti-Human, Mouse, Clone: OTI4D12, lyophilized, TrueMAB™

Codice prodotto. 17653881
Encompass_Preferred
Cambia vista
Click to view available options
Quantità:
100 μg
Dimensione della confezione:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
17653881 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 17653881 Fornitore Origene Technologies N. del fornitore CF504436

Please to purchase this item. Need a web account? Register with us today!

Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.

Richiedi qui il prezzo per questo articolo.

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the proteasome B-type family, also known as the T1B family, that is a 20S core beta subunit in the proteasome. Expression of this catalytic subunit is downregulated by gamma interferon and proteolytic processing is required to generate a mature subunit. This subunit is not present in the immunoproteasome and is replaced by catalytic subunit 2i (proteasome beta 10 subunit).

Specifica

Antigene PSMB7
Applicazioni Western Blot
Classificazione Monoclonal
Clone OTI4D12
Coniugato Unconjugated
Formulazione PBS with 8% trehalose and no preservative; pH 7.3
Gene PSMB7
N. accesso geni P70195, Q99436
Alias gene Z
Simboli geni Psmb7
Specie ospite Mouse
Immunogeno Human recombit protein fragment corresponding to amino acids 58-277 of human PSMB7 produced in E.coli.
Metodo di purificazione Affinity Chromatography
Quantità 100 μg
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 19177, 5695
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Forma Lyophilized
Isotype IgG1
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.