missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMB8 Mouse anti-Human, Rat, Clone: 2A5B6, Proteintech
Mouse Monoclonal Antibody
140.00€ - 399.00€
Specifica
Antigene | PSMB8 |
---|---|
Clone | 2A5B6 |
Concentrazione | 1 mg/mL |
Applicazioni | Western Blot, Immunohistochemistry (Paraffin) |
Classificazione | Monoclonal |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
16808935
|
Proteintech
66759-1-IG-20UL |
20 μL |
140.00€
20µL |
Spedizione stimata: 10-05-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
16898925
|
Proteintech
66759-1-IG-150UL |
150 μL |
399.00€
150µL |
Spedizione stimata: 10-05-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Descrizione
The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the proteasome B-type family, also known as the T1B family, that is a 20S core beta subunit. This gene is located in the class II region of the MHC (major histocompatibility complex). Expression of this gene is induced by gamma interferon and this gene product replaces catalytic subunit 3 (proteasome beta 5 subunit) in the immunoproteasome. Proteolytic processing is required to generate a mature subunit. Two alternative transcripts encoding two isoforms have been identified; both isoforms are processed to yield the same mature subunit.Specifica
PSMB8 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
beta5i, D6S216, D6S216E, LMP7, Low molecular mass protein 7, Macropain subunit C13, Proteasome component C13, Proteasome subunit beta 5i, Proteasome subunit beta type 8, PSMB5i, PSMB8, RING10, Y2 | |
PSMB8 | |
IgG1 | |
Protein G | |
Antibody |
2A5B6 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Rat, Human | |
P28062, P28064 | |
24968, 5696 | |
PSMB8 Fusion Protein Ag6780 | |
Primary | |
-20°C | |
PSMB8 |