Learn More
Horst™ Pt 100 Sensor
Product Code. 10719822
Change view
Click to view available options
Diameter (Metric):
4mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
10719822 4mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10719822 Supplier Horst™ Supplier No. 70308
  • ONLINE EXCLUSIVE PRICE
    155.44€
    96.33€ / pezzi 1
    Risparmia 59.11€
    38% Sconto
 Restricted Item
Spedizione stimata: 02-12-2026
Specifications
Diameter (Metric) 4mm
Diameter (Metric) Inner 4 mm
Length (Metric) 50 mm
Material Fiber Glass
Length (Metric) Power Cord 50mm
Product Type Pt 100 Sensor
Temperature (Metric) 500°C