Learn More
Masterflex™ PTFE 3-Way Direct Lift Solenoid Valve
Product Code. 12843223 Shop All Masterflex Products
Change view
Click to view available options
Voltage:
12 VDC
24 VDC
Maximum Pressure:
18 psi
30 psi
60 psi
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Maximum Pressure Unit Size
12843223 24 VDC 30 psi Each
11700029 12 VDC 30 psi Each
11790019 24 VDC 60 psi Each
13257499 24 VDC 18 psi Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12843223 Supplier Masterflex™ Supplier No. 0154012
  • ONLINE EXCLUSIVE PRICE
    273.00€
    193.97€ / pezzi 1
    Risparmia 79.03€
    28% Sconto
Spedizione stimata: 23-12-2026
Specifications
For Use With (Application) Ideal for use with high-purity or corrosive fluids
Max. Operating Temperature (Metric) 70°C
Material PTFE
Quantity 1 Each
Type 3-way Direct Lift Solenoid Valve
Voltage 24 VDC
Maximum Pressure 30 psi