Learn More
Aalborg™ PTFE Aluminum Shell Needle Valve
Codice prodotto. 11860120
Cambia vista
Click to view available options
Include:
1/8 in. NPTF Fitting
Glass Nipple Fitting
Dimensione della confezione:
Pezzo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Include unitSize
11860120 1/8 in. NPTF Fitting Pezzo
11820140 Glass Nipple Fitting Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 11860120 Fornitore Aalborg™ N. del fornitore VCHTT0A
Please to purchase this item. Need a web account? Register with us today!
Aalborg™ PTFE Aluminum Shell Needle Valve is designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
  • Fluids in contact with PTFE only
  • Structurally rigid metal shell.
  • One PTFE o-ring
  • Simplicity - only seven components.
  • Up to 55 SLPM (air) or 2.8 LPM (water)
Specifica
Tipo Needle Valve
Include 1/8 in. NPTF Fitting
Velocità di flusso dell'aria 55 L/min.
Portata flusso d'acqua 28 mL/min.
Collegamento ingresso NPTF (sistema imperiale) 1/8 in. NPTF
Temperatura operativa max (sistema metrico) 65°C