missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PTFE Diagnostic Slides

These hydrophobic coatings bind liquids applied to the wells and keeps fluids in the well area. Clean, flat and suitable for immediate use

Marca:  Thermo Scientific™ ER285W

Dettagli aggiuntivi : Peso : 1.00000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15394898

  • 87.24€ / pezzi 3000


Per tempo di consegna, consultarci allo 02 950 59 478
Aggiungi al carrello

Descrizione

Descrizione

  • Binds and keeps liquids applied to the well area
  • PTFE Construction
  • Provides wettable wells for staining and growing cell cultures
  • Prevents cross-contamination
  • Hydrophobic coatings are very durable and withstand the strongest stains
  • Autoclavable
Specifica

Specifica

25mm
White
PTFE
100/PK
1
75mm
90°, ground edge
Diagnostic Slide
24.4 x 16.7mm Oval
Videos