Learn More
Abnova™ PTP4A3/CEN8q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Marca:  Abnova™ FG0170.100uL
Dettagli aggiuntivi : Peso : 0.02000kg
Codice prodotto. 16013265
  • 1285.26€ / 100µL
Spedizione stimata
07-06-2021
Specifica
FISH-Ce
Liquid
100 µL
11156
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Store at 4°C in the dark.