Learn More
Saint-Gobain Performance Plastics PureFlo™ PTFE Disc Capsules
Description
- PureFlo™ PTFE Capsules are designed for single-use filtration applications and provide sterility and convenience in pharmaceutical processes.
- The hydrophobic PTFE membrane delivers reliable performance in venting and compressed gas applications.
- The capsules are autoclavable and meet stringent sterilization requirements for pharmaceutical use.
- The sterilizing grade 0.2 μm membrane ensures superior flow efficiency and reduces pressure drops compared to other membranes like PVDF.
- Animal-derived components are excluded to meet industry safety standards and compliance requirements.
- The capsules offer robust chemical resistance, making them ideal for use in demanding environments.
- Each capsule undergoes rigorous integrity testing to guarantee consistent and dependable performance.
- The compact and lightweight design allows easy integration into various assemblies without adding bulk.
- A versatile size range supports scale-up efforts throughout all stages of drug development and manufacturing processes.
Specifications
Specifications
| Autoclavabile | Not Autoclavable |
| Monouso | Single-use |
| Tipo di filtro | Filter Capsule |
| Materiale (membrana) | Hydrophilic PE |
| Dimensione del poro | 0.2 μm |
| Sterilità | Non-sterile |
| Tipo | Carboy Vent |
| Diametro (sistema metrico) | 33 mm Body |