Learn More
PURG Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA561583
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: RGGKNVGGSG LSKSRLYPQA QHSHYPHYAA SATPNQAGGA AEIQELASK Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 92%.
The exact function of this gene is not known, however, its encoded product is highly similar to purine-rich element binding protein A. The latter is a DNA-binding protein which binds preferentially to the single strand of the purine-rich element termed PUR, and has been implicated in the control of both DNA replication and transcription. PURG lies in close proximity to the Werner syndrome gene, but on the opposite strand, on chromosome 8p11. Two transcript variants encoding different isoforms have been found for this gene.Specifica
PURG | |
Polyclonal | |
Unconjugated | |
PURG | |
4930486B15Rik; MGC119274; Purg; PURG-A; Pur-gamma; PURG-B; purine rich element binding protein G; purine-rich element binding protein G; Purine-rich element-binding protein G; Purine-rich element-binding protein gamma | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29942 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UJV8 | |
PURG | |
Recombinant protein corresponding to Human PURG | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only