Learn More
Masterflex™ PVDF Barbed NPT Male Pipe Adapter Elbows
Designed for quick and easy assembly without tools
Marca:  Masterflex™ 30704-43
Dettagli aggiuntivi : Peso : 0.80000kg
Codice prodotto. 11846743
  • 56.51€ / pezzi 10
Spedizione stimata
02-07-2021
Aggiungi al carrello
Descrizione
Descrizione
  • Ideal for low-pressure applications
  • Chemical resistant
  • Available in a variety of sizes
  • Economical barbed fittings easily connect fluid paths without the use of tools
  • Just slip your flexible tubing or hose onto the barb to create a secure seal
Specifica
Specifica
Translucent
Autoclavable
1 27/32 in.
1/2 in.
1/2 in.
Autoclave or Ethylene Oxide
1 29/32 in.
1/2 NPT(M) x 1/2 in. ID
PVDF
Barbed NPT Male Pipe Adapter Elbows
1/2 in.
1/2 in.
1/2 in.