Learn More
Masterflex™ PVDF NPT Male Adapter to Hose Barb
Connect fluid paths without the use of tools
Marca:  Masterflex™ 30704-20
Dettagli aggiuntivi : Peso : 0.80000kg
Codice prodotto. 11836703
  • 53.85€ / pezzi 10
Spedizione stimata
29-06-2021
Aggiungi al carrello
Descrizione
Descrizione
  • Quick and easy assembly without tools
  • Just slip your flexible tubing or hose onto the barb to create a secure seal
  • Chemical resistant
  • Available in a variety of sizes
Specifica
Specifica
Translucent
Autoclavable
2 1/4 in.
3/4 in.
3/4 in.
Autoclave or Ethylene Oxide
1 in.
Barb x NPT(M) Connection
PVDF
NPT Male Adapter to Hose Barb
3/4 in.
1/2 in.
1/2 in.