Descrizione
These straight-through valves with electromagnetic (solenoid) drive are used for operating cycles with short intervals and, therefore, are often used as vacuum control valves. The straightforward design provides a valve with low leakage rate. The valves in version VV C are made of materials with excellent chemical resistance. The valve sealing of the VV-B 6C is made of a special fluoroelastomer with better form stability than common PTfE, much higher chemical resistance than fKM and excellent long-term leak tightness characteristics.
- version C with excellent chemical resistance
- high operating cycles
- significantly better long term tightness even after long operating time
- easy to clean
- versatile connection alternatives via small flange or hose nozzle
Specifica
Specifica
| Materiale | PVDF, PTFE |
| Quantità | 1 Each |
| Tipo | In-Line Isolation Valve |