missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Purity: >98%
- Molecular Weight: 210.23 Da
- Biological Activity: Catalase inhibitor, aryl hydrocarbon receptor activator (Ki = 5.4 μM), generates ROS
- Biological Effects: Reduces gene expression of catalase, promotes extracellular DNA release, and induces apoptosis in neutrophils in vivo
- Form: Solid
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Tampone | 100% Pyocyanin |
| Content And Storage | Store at -20°C. |
| Forma | Solid |
| Shipping Condition | Ambient |
| Target Molecule | Catalase, Aryl Hydrocarbon Receptor |
| Formulazione | 100% Pyocyanin |
| Quantità | 5 mg |
| Da utilizzare con (applicazione) | Catalase inhibition, Aryl hydrocarbon receptor activation, ROS generation, Apoptosis induction |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?