Learn More
Pyrex™ Borosilicate Glass Cylindrical Reaction Vessel with Thermostat Valve
Specifica
Specifica
| Capacità (sistema metrico) | 2000 mL |
| Materiale | Borosilicate Glass |
| Altezza (sistema metrico) | 340 mm |
| Product Type | Reaction vessel |
| Forma | Cylindrical |