Learn More
Pyrex™ Borosilicate Glass Hollow Blown Stopper
Hollow-blown Quickfit™ jointed glassware featuring C length ground glass sockets.
Marca:  Quickfit 2595/07M
235.60 EUR valido fino al 2024-12-31
Usa il codice promo "23259" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.60000kg
Codice prodotto. 12437017
  • 248.00€ / pezzi 20
Spedizione stimata: 10-07-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Ideally suited for use with Quickfit™ jointed glassware and other glassware having the shorter C length ground glass sockets
  • All Quickfit™ stoppers conform to the standard tapers as outlined in ISO 383
  • Borosilicate glass for chemical durability and high temperature performance
  • Hollow-blown construction to reduce weight
  • Hexagonal head prevents the stopper from rolling and provides a good grip
  • Top of stopper is flat or slightly concave, allowing them to be stood on end and avoid contamination of the ground surface
Specifica
Specifica
Cone size: 29/22
68mm
Borosilicate Glass
ISO 383