missing translation for 'onlineSavingsMsg'
Learn More

Microbiologics™ QA/QC Microorganism Preparations in KWIK-STIK™ Plus

Product Code. 11961406
Encompass
Change view
Click to view available options
Species:
Aspergillus brasiliensis
Bacillus subtilis
Escherichia coli
Salmonella typhimurium
ATCC®:
6633
8739
14028
16404
Biosafety Level:
1
2
Unit Size:
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Species ATCC® Biosafety Level Unit Size
11961406 Escherichia coli 8739 2 Pack of 5
11906635 Salmonella typhimurium 14028 2 Pack of 5
11951426 Bacillus subtilis 6633 2 Pack of 5
11946675 Aspergillus brasiliensis 16404 1 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11961406 Supplier Microbiologics™ Supplier No. 0483X
 Restricted Item
Spedizione stimata: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Come in canisters of five units of a single strain

†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.

‡

Biotype G

Specifications

ATCC® 8739
Biosafety Level 2
Format KWIK-STIK™ Plus
Product Type KWIK-STIK
Packaging Type Canister
Quantity 5 units of a single strain
Species Escherichia coli
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.